missing translation for 'onlineSavingsMsg'
Learn More

CoraLite™ Plus 488-conjugated RAB7A Rabbit Polyclonal Antibody, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech CL488-55469-100UL

 View more versions of this product

Product Code. 30234552

  • € 450.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RAB7A antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human RAB7A. This antibody recognizes human, mouse, rat antigen. The RAB7A antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 488.
Specifications

Specifications

RAB7A
Polyclonal
CoraLite Plus 488
Rabbit
Antigen affinity purification
RUO
7879
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Conjugated Antibody
IgG
Flow Cytometry (Intracellular Staining)
1000 μg/mL
FLJ20819; PRO2706; RAB7
Peptide
100 μL
Primary
Human, Mouse, Rat
23 kDa
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.