missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CEACAM1/CEACAM3/CEACAM6 Monoclonal Antibody (YTH71.3)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA190031
This item is not returnable.
View return policy
Description
MA1-90031 detects CD66a, CD66c + CD66d from non-human primate, human samples.
This gene encodes a member of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily. Two subgroups of the CEA family, the CEA cell adhesion molecules and the pregnancy-specific glycoproteins, are located within a 1.2 Mb cluster on the long arm of chromosome 19. Eleven pseudogenes of the CEA cell adhesion molecule subgroup are also found in the cluster. The encoded protein was originally described in bile ducts of liver as biliary glycoprotein. Subsequently, it was found to be a cell-cell adhesion molecule detected on leukocytes, epithelia, and endothelia. The encoded protein mediates cell adhesion via homophilic as well as heterophilic binding to other proteins of the subgroup. Multiple cellular activities have been attributed to the encoded protein, including roles in the differentiation and arrangement of tissue three-dimensional structure, angiogenesis, apoptosis, tumor suppression, metastasis, and the modulation of innate and adaptive immune responses. Multiple transcript variants encoding different isoforms have been reported, but the full-length nature of all variants has not been defined.
Specifications
| CEACAM1/CEACAM3/CEACAM6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| P13688, P40198, P40199 | |
| CEACAM1, CEACAM3, CEACAM6 | |
| Human lymphocytes. | |
| 100 μg | |
| Primary | |
| Human, Rhesus Macaque | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| YTH71.3 | |
| Unconjugated | |
| CEACAM1 | |
| antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen CGM1; carcinoembryonic antigen CGM6; carcinoembryonic antigen gene family member 1; carcinoembryonic antigen gene family member 6; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 3; carcinoembryonic antigen related cell adhesion molecule 6; carcinoembryonic antigen related cell adhesion molecule 8; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 3; Carcinoembryonic antigen-related cell adhesion molecule 6; carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific cross reacting antigen); carcinoembryonic antigen-related cell adhesion molecule 8; CD66a; CD66a antigen; CD66b; CD66c; CD66D; CD66d antigen; CD67; CD67 antigen; CEA; CEA cell adhesion molecule 1; CEA cell adhesion molecule 3; CEA cell adhesion molecule 6; CEACAM1; CEACAM3; CEACAM6; CEACAM8; CEAL; CGM1; CGM6; Cluster of Differentiation 66c; NCA; NCA-95; Non-specific crossreacting antigen; nonspecific cross-reacting antigen; non-specific cross-reacting antigen NCA-95; normal cross-reacting antigen; pan CEACAM; W264; W282 | |
| Rat | |
| Protein G | |
| RUO | |
| 106994775, 1084, 4680, 634, 707824, 708200 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction