missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CEACAM1/CEACAM5 Recombinant Rabbit Monoclonal Antibody (BLR032F)
GREENER_CHOICE

Product Code. 17755629
Change view
Click to view available options
Quantity:
10 μL
100 μL
Unit Size:
100µL
10µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17755629 100 μL 100µL
17765629 10 μL 10µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17755629 Supplier Invitrogen™ Supplier No. MA544259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

Over-expression of CEA/CEACAM5 in tumors of epithelial origin is the basis of its wide-spread use as a tumor marker CEACAM1 expression is down-regulated in many tumors indicating a tumor-suppressive function. The anti-tumor effect may be due to inhibition of tumor angiogenesis, possibly by increased secretion of anti-angiogenic molecules from the cells. The function of CEA family members varies widely: they function as cell adhesion molecules, tumor suppressors, regulators of lymphocyte and dendritic cell activation, receptors of Neisseria species and other bacteria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM1/CEACAM5
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR032F
Concentration 50 μg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene CEACAM1
Gene Accession No. P06731, P13688
Gene Alias antigen CD66; BGP; BGP1; BGP-1; BGPI; Biliary glycoprotein 1; Carcinoembryonic antigen; carcinoembryonic antigen related cell adhesion molecule 1; carcinoembryonic antigen related cell adhesion molecule 5; carcinoembryonic antigen-related cell adhesion molecule 1; carcinoembryonic antigen-related cell adhesion molecule 1 (biliary glycoprotein); carcinoembryonic antigen-related cell adhesion molecule 5; CD66a; CD66a antigen; CD66e; CEA; CEACAM1; CEACAM5; Meconium antigen 100; pan CEACAM; sCD66e; soluble CD66e
Gene Symbols CEACAM1, CEACAM5
Host Species Rabbit
Immunogen Recombinant protein representing the extracellular domain of human CEACAM1 (residues 35-433).
Purification Method Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1048, 634
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.