missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM19 Antibody (1032215), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9397H
This item is not returnable.
View return policy
Description
CEACAM19 Monoclonal antibody specifically detects CEACAM19 in Human samples. It is validated for Flow Cytometry
Specifications
| CEACAM19 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Mouse | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 56971 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| 1032215 | |
| Flow Cytometry | |
| carcinoembryonic antigen-related cell adhesion molecule 19 | |
| Human embryonic kidney cell HEK293-derived human CEACAM-19, Ala33-Gly157, Accession # Q7692-1 | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction