missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM19 Antibody (1042108), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB93971MFV450
This item is not returnable.
View return policy
Description
CEACAM19 Monoclonal antibody specifically detects CEACAM19 in Human samples. It is validated for Flow Cytometry
Specifications
| CEACAM19 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1042108 | |
| Flow Cytometry | |
| carcinoembryonic antigen-related cell adhesion molecule 19 | |
| Chinese hamster ovary cell line CHO-derived recombinant human CEACAM-19, Met1-Gly157, Accession # Q7Z692 | |
| 0.1 mL | |
| Apoptosis, Cancer | |
| 56971 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction