Learn More
CEACAM2 Antibody (935610) [FITC], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB9214F
Code produit. 18611834
  • € 610.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
CEACAM2 Monoclonal antibody specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Spécification
Spécification
CEACAM2
Monoclonal
FITC
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
26367
Store at 4°C in the dark.
IgG1
Flow Cytometry, CyTOF
935610
Flow Cytometry, CyTOF-ready
Bgp;Bgp2;Carcinoembryonic antigen-related cell adhesion molecule 2;Ceacam2-L
Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2, Gln35-Asp422, Accession # Q925P2
0.1 mL