missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM2 Antibody (935610) [Janelia Fluor™ 525], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB9214JF525
This item is not returnable.
View return policy
Description
CEACAM2 Monoclonal antibody specifically detects CEACAM2 in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CEACAM2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Bgp;Bgp2;Carcinoembryonic antigen-related cell adhesion molecule 2;Ceacam2-L | |
| Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2, Gln35-Asp422, Accession # Q925P2 | |
| RUO | |
| 26367 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 935610 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction