Learn More
Invitrogen™ CEACAM2 Monoclonal Antibody (935610)
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  Invitrogen™ MA562008
Product Code. 30535124
  • € 449.00 / 100µg
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
CEACAM2 (Carcinoembryonic Antigen-Related Cell Adhesion Molecule 2) is a gene that encodes a protein belonging to the carcinoembryonic antigen (CEA) family, which is part of the immunoglobulin superfamily. CEACAM2 is involved in various cellular processes, including cell adhesion, intracellular signaling, and immune responses. It is expressed in several tissues, including the brain, liver, and epithelial cells of the gastrointestinal tract. CEACAM2 plays a role in maintaining tissue architecture and integrity by mediating cell-cell adhesion. It also participates in immune regulation by interacting with other cell surface molecules to modulate the activity of immune cells, such as macrophages and natural killer (NK) cells. Dysregulation or altered expression of CEACAM2 has been associated with various diseases, including cancer, where it can influence tumor progression and metastasis, and metabolic disorders, such as obesity and type 2 diabetes. Understanding the functions and mechanisms of CEACAM2 is crucial for developing therapeutic strategies aimed at targeting its role in disease processes. Research into CEACAM2 offers insights into its potential as a biomarker for disease diagnosis and prognosis, as well as a target for novel treatments in oncology and immunotherapy.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEACAM2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Mouse myeloma cell line NS0-derived recombinant mouse CEACAM-2, Gln35-Asp422.
100 μg
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Flow Cytometry
935610
Unconjugated
Rat
Protein A/G