missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM3/CD66d Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99909-50ul
This item is not returnable.
View return policy
Description
CEACAM3/CD66d Polyclonal antibody specifically detects CEACAM3/CD66d in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| CEACAM3/CD66d | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| CD66D, CEA, CEACAM3 carcinoembryonic antigen-related cell adhesion molecule 3, CGM1, W264, W282 | |
| Produced in rabbits immunized with purified, recombinant Human CEACAM3/CD66d (Accession#: NP_001806.2; Met1-Gly155) | |
| 50 μg | |
| CD Markers, Immunology, Innate Immunity | |
| 1084 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction