missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CD66e Antibody (061), Biotin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89785B
This item is not returnable.
View return policy
Description
CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry.
Specifications
| CEACAM5/CD66e | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 61 | |
| Flow Cytometry | |
| Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM5/CD66e (NP_004354.2; Met1-Ala685). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Immunology | |
| 1048 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction