missing translation for 'onlineSavingsMsg'
Learn More

CEACAM5/CD66e Antibody (308/3-3), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-54624AF750

Product Code. 18240305

  • ONLINE EXCLUSIVE PRICE
    € 488.00
    € 460.95 / 0.10mL
    Save € 27.05
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Flow Cytometry, Sandwich ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CEACAM5/CD66e
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
CEACAM5
Recombinant soluble human CEACAM3-Fc produced in HEK293 cells
0.1 mL
1048
Human
IgG1 κ
Western Blot, Flow Cytometry, ELISA
308/3-3
Western Blot, Flow Cytometry, ELISA, Sandwich ELISA
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
Protein A purified
Primary
Clone 308/3-3 reacts with human CEACAM3 and CEACAM5, but not human CEACAM1/4/6/7/8, rat CEACAM1, or mouse CEACAM1 & CEACAM2.
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.