missing translation for 'onlineSavingsMsg'
Learn More

CEACAM5/CD66e Antibody (308/3-3), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-54624PEATT594

Product Code. 18220484

  • € 487.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CEACAM5/CD66e Monoclonal specifically detects CEACAM5/CD66e in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CEACAM5/CD66e
Monoclonal
PE/ATTO 594
PBS with 0.05% Sodium Azide
CEACAM5
Recombinant soluble human CEACAM3-Fc produced in HEK293 cells
0.1 mL
Primary
Clone 308/3-3 reacts with human CEACAM3 and CEACAM5, but not human CEACAM1/4/6/7/8, rat CEACAM1, or mouse CEACAM1 & CEACAM2.
Store at 4°C in the dark. Do not freeze.
Flow Cytometry
308/3-3
Flow Cytometry
Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100
Mouse
Protein A purified
Apoptosis, Cancer, Cellular Markers, Immunology
1048.0
Human
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.