missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5/CD66e Antibody (5C8C4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54625APC
This item is not returnable.
View return policy
Description
CEACAM5/CD66e Monoclonal antibody specifically detects CEACAM5/CD66e in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOF
Specifications
| CEACAM5/CD66e | |
| Monoclonal | |
| APC | |
| Carcinoembryonic antigen, carcinoembryonic antigen-related cell adhesion molecule 5, CD66e antigen, CEACD66e, DKFZp781M2392, Meconium antigen 100 | |
| Soluble human CEACAM5/CD66e | |
| 0.1 mL | |
| Cancer, Cellular Markers, Immunology | |
| 1048 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 5C8C4 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction