missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM6/CD66c Rabbit anti-Human, Clone: 408, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 370.65 - € 546.00
Specifications
| Antigen | CEACAM6/CD66c |
|---|---|
| Clone | 408 |
| Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:100-1:500 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18221428
|
Novus Biologicals
NBP2-89669-50UL |
50 μg |
€ 392.00 € 370.65 / 50µL Save € 21.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18261966
|
Novus Biologicals
NBP2-89669-100UL |
100 μg |
€ 578.00 € 546.00 / 100µL Save € 32.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CEACAM6/CD66c Monoclonal specifically detects CEACAM6/CD66c in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| CEACAM6/CD66c | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:100-1:500, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| carcinoembryonic antigen-related cell adhesion molecule 6, carcinoembryonic antigen-related cell adhesion molecule 6 (non-specific crossreacting antigen), CD66c, CD66c antigen, NCACEAL, Non-specific crossreacting antigen, Normal cross-reacting antigen | |
| Recombinant Human CEACAM6/CD66c protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 408 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 0.2 um filtered solution in PBS | |
| 4680 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title