Learn More
CEACAM7 Antibody (962720), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB44782MFV450
This item is not returnable.
View return policy
Description
CEACAM7 Monoclonal antibody specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
Specifications
| CEACAM7 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Cancer | |
| 1087 | |
| Store at 4C in the dark. |
| Flow Cytometry, ELISA, Immunohistochemistry, CyTOF | |
| 962720 | |
| Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready | |
| Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CGM 2, CGM2 | |
| Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7, Thr36-Asn233, Accession # Q14002 | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |