Learn More
CEACAM7 Antibody (962720), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB44782MFV450
Product Code. 18597507
  • € 636.00 / 0.10mL
Description
CEACAM7 Monoclonal antibody specifically detects CEACAM7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CEACAM7
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
0.1 mL
Cancer
1087
Store at 4C in the dark.
Flow Cytometry, ELISA, Immunohistochemistry, CyTOF
962720
Flow Cytometry, ELISA, Immunohistochemistry, CyTOF-ready
Carcinoembryonic antigen CGM 2, Carcinoembryonic antigen CGM2, Carcinoembryonic antigen gene family member 2, Carcinoembryonic antigen related cell adhesion molecule 7, CEA, CEACAM 7, CGM 2, CGM2
Chinese Hamster Ovary cell line, CHO-derived recombinant human CEACAM-7, Thr36-Asn233, Accession # Q14002