missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM8/CD66b Antibody (6/40c), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54627R
This item is not returnable.
View return policy
Description
CEACAM8/CD66b Monoclonal antibody specifically detects CEACAM8/CD66b in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, CyTOF
Specifications
| CEACAM8/CD66b | |
| Monoclonal | |
| DyLight 550 | |
| Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95 | |
| Recombinant soluble human CEACAM8-Fc produced in HEK293 cells | |
| 0.1 mL | |
| Cancer | |
| 1088 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| 6/40c | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction