missing translation for 'onlineSavingsMsg'
Learn More

CEACAM8/CD66b Antibody (G10F5), Novus Biologicals™

Product Code. 18596963 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18596963 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18596963 Supplier Novus Biologicals Supplier No. NB10077808SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

CEACAM8/CD66b Monoclonal specifically detects CEACAM8/CD66b in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunocytochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CEACAM8/CD66b
Applications Flow Cytometry
Classification Monoclonal
Clone G10F5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1 μg/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33122293), Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry, CyTOF-ready
Gene Accession No. P31997
Gene Alias Carcinoembryonic antigen CGM6, carcinoembryonic antigen gene family member 6, carcinoembryonic antigen-related cell adhesion molecule 8, CD66b, CD66b antigen, cd66b clone g10f5, cd66b G10F5, cd66b granulocyte marker, cd66b ihc, cd66b marker, cd66b neutrophil marker, cd66b neutrophils, CD67, CD67 antigen, CGM6, NCA-95, Non-specific cross-reacting antigen NCA-95
Gene Symbols CEACAM8
Host Species Mouse
Immunogen Human peripheral blood cells.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 1088
Test Specificity CEACAM8 (G10F5). Expected Cross-Reactivity based on immunogen sequence: Chimpanzee.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.