missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBP Beta Antibody (47A1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04266JF646
This item is not returnable.
View return policy
Description
CEBP Beta Monoclonal antibody specifically detects CEBP Beta in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| CEBP Beta | |
| Monoclonal | |
| Janelia Fluor 646 | |
| C/EBP beta, C/EBP-beta, CCAAT/enhancer binding protein (C/EBP), beta, CCAAT/enhancer-binding protein beta, CRP2, IL6DBP, interleukin 6-dependent DNA-binding protein, LAPTCF-5, Liver activator protein, liver-enriched transcriptional activator protein, MGC32080, NF-IL6, Nuclear factor NF-IL6, nuclear factor of interleukin 6, TCF5NFIL6, Transcription factor 5 | |
| Recombinant human CEBP Beta (1-271 aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| 47A1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 1051 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction