Learn More
CEBPA Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 29388-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The CEBPA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CEBPA. This antibody recognizes human antigen. The CEBPA antibody has been validated for the following applications: IP, WB, ELISA analysis.
Spécification
| CEBPA | |
| 1500 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunoprecipitation, ELISA | |
| C/EBP alpha, CEBP, CEBPA | |
| 150 μL | |
| 1050 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |