missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEBPB Mouse anti-Human, Rat, Clone: 2B6E10, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | CEBPB |
|---|---|
| Clone | 2B6E10 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
16848725
|
Proteintech
66649-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16838725
|
Proteintech
66649-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct DNA-binding forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the development of mesenchymal tissues.Spécification
| CEBPB | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| C/EBP beta, CEBPB, CRP2, IL6DBP, LAP, Liver activator protein, NF IL6, Nuclear factor NF IL6, TCF 5, TCF5, Transcription factor 5 | |
| CEBPB | |
| IgG1 | |
| Protein G | |
| Antibody |
| 2B6E10 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P17676, P21272 | |
| 1051, 24253 | |
| CEBPB Fusion Protein Ag20073 | |
| Primary | |
| -20°C | |
| CEBPB |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit