Learn More
Cefoperazone Acid, TRC
Specifications
Specifications
| Purity | >95% (HPLC) |
| Format | Neat |
| Molecular Weight (g/mol) | 645.67 |
| Product Type | Metabolite∣API |
| Quantity | 1 g |
| Shipping Condition | Room Temperature |