Learn More
Thermo Scientific Chemicals Cefoxitin sodium salt, 94%
Catalog Number p-7077540
Click to view available options
Quantity:
1 g
5 g
33564-30-6
C16H16N3NaO7S2
449.428
GNWUOVJNSFPWDD-XMZRARIVSA-M
23667300
CHEBI:3501
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
33564-30-6
C16H16N3NaO7S2
449.428
GNWUOVJNSFPWDD-XMZRARIVSA-M
23667300
CHEBI:3501
sodium;(6R,7S)-3-(carbamoyloxymethyl)-7-methoxy-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
COC1(C2N(C1=O)C(=C(CS2)COC(=O)N)C(=O)[O-])NC(=O)CC3=CC=CS3.[Na+]
Cefoxitin sodium salt, CAS # 33564-30-6, is a β-lactam, second-generation cephalosporin compound showing bactericidal activity against Bacteroides, gram-negative bacteria, including strains of P. mirabilis, P. vulgaris, E. coli, S. typhimurium, and S. enteritidis.
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Cefoxitin sodium salt is resistant to β-lactamase
• It binds and inactivates penicillin-binding proteins (PBP) located on the inner membrane of the bacterial cell wall thus interfering with the cross-linkage of peptidoglycan chains necessary for bacterial cell wall strength and rigidity
Applications
• Cefoxitin sodium salt is the sodium salt form of cefoxitin derived from cephamycin C
• It is a β-lactam, second-generation cephalosporin compound with bactericidal activity against numerous species such as Bacteroides, gram-negative bacteria, including strains of P. mirabilis, P. vulgaris, E. coli, S. typhimurium, and S. enteritidis
• Cefoxitin increases survival in mice infected with S. aureus, S. pyogenes, and K. pneumoniae
TRUSTED_SUSTAINABILITY
Type Cefoxitin Sodium Salt
Infrared Spectrum Authentic
Formula Weight 449.43
Specific Rotation +206° to +214° (20°C, 589nm) (c= 1, methanol)
Percent Purity 94%
Water 1% max.
Quantity 1 g
Chemical Name or Material Cefoxitin sodium salt
compliance-icons
Product Identifier
  • Cefoxitin sodium salt
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.