missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime pentahydrate, 98%, ACROS Organics™

Brand:  Acros Organics 461730250

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 15741767

  • € 177.00 / 25g


Estimated Shipment
11-12-2020
Add to basket

Description

Description

Specifications

Chemical Identifiers

78439-06-2
636.65
131673973
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
C22H22N6O7S2·5H2O
NMVPEQXCMGEDNH-TZVUEUGBSA-N
CHEBI:3509
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O

Specifications

Ceftazidime pentahydrate
25g
97.5% min. (HPLC)
13,1959
NMVPEQXCMGEDNH-TZVUEUGBSA-N
2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
131673973
636.65
>150.0°C
78439-06-2
C22H22N6O7S2·5H2O
Solubility in water: slightly soluble. Other solubilities: soluble in alkali,slightly soluble in methanol,dmf,insoluble in acetone,ethanol,chloroform,ether,,ethyl acetate,toluene,dioxane
CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O
636.65
CHEBI:3509
98%
Videos
SDS
Documents