missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CELA1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78085PE
This item is not returnable.
View return policy
Description
CELA1 Polyclonal antibody specifically detects CELA1 in Mouse, Porcine samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| CELA1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Primary | |
| Mouse, Pig | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| PE | |
| chymotrypsin-like elastase family member 1, chymotrypsin-like elastase family, member 1, EC 3.4.21, EC 3.4.21.36, ELA1elastase-1, elastase 1, pancreatic, Elastase-1 | |
| CELA1 [Porcine Pancreas] (Uniprot: P00772) | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Lipid and Metabolism | |
| 1990 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction