missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CELA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122103
This item is not returnable.
View return policy
Description
CELA1 Polyclonal Antibody for Western Blot, IHC (P)
Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, pancreatic elastase 1 is not expressed in the pancreas. To date, elastase 1 (ELA1) expression has only been detected in skin keratinocytes. Clinical literature that describes human elastase 1 activity in the pancreas is actually referring to elastase 2A.Elastases form a subfamily of serine proteases that hydrolyze many proteins in addition to elastin. Humans have six elastase genes which encode the structurally similar proteins elastase 1, 2, 2A, 2B, 3A, and 3B. Unlike other elastases, pancreatic elastase 1 is not expressed in the pancreas. To date, elastase 1 expression has only been detected in skin keratinocytes. Clinical literature that describes human elastase 1 activity in the pancreas is actually referring to elastase 2A.
Specifications
| CELA1 | |
| Polyclonal | |
| Unconjugated | |
| CELA1 | |
| 1810009A17Rik; 1810062B19Rik; Cela1; chymotrypsin like elastase 1; chymotrypsin like elastase family member 1; chymotrypsin-like elastase family member 1; chymotrypsin-like elastase family, member 1; ELA1; Ela-1; ELAI1; elastase 1; elastase 1, pancreatic; elastase I; Elastase-1; ELS1; pancreatic elastase 1; pancreatic elastase I; pancreatic preproelastase I; PC-TsF; preproelastase I; RATELAI1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 109901, 24331, 396766 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P00772, P00773, Q91X79 | |
| CELA1 | |
| Recombinant protein Pancreatic Elastase 1. The antigen corresponds to amino acid range 44-259 of the target protein. | |
| 20 μL | |
| Primary | |
| Mouse, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction