missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CELA3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115171
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Elastase.
Efficient protease with alanine specificity but only little elastolytic activity.
Specifications
| CELA3B | |
| Polyclonal | |
| Unconjugated | |
| CELA3B | |
| 0910001F22Rik; 2310074F01Rik; AI504000; CBPP; Cela3b; cholesterol-binding pancreatic protease; chymotrypsin like elastase family member 3B; chymotrypsin-like elastase family member 3B; chymotrypsin-like elastase family, member 3B; Ela3; ELA3B; elastase 3, pancreatic; elastase 3B, pancreatic; elastase IIIB; Elastase-3B; fecal elastase 1; pancreatic elastase 1; pancreatic endopeptidase E; procarboxypeptidase A complex component III; procarboxypeptidase A-S6 subunit III; PROCPA-S6 III; proproteinase E; protease E; proteinase E | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23436, 298567, 67868 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P08861, Q9CQ52 | |
| CELA3B | |
| A synthesized peptide derived from human CELA3B(Accession P08861), corresponding to amino acid residues V68-V118. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction