missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CELA3B Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
€ 185.00 - € 393.00
Specifications
| Antigen | CELA3B |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17900233
|
Invitrogen™
PA5145364 |
20 μL |
€ 185.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17940853
![]() |
Invitrogen™
PA5145363 |
200 μL |
€ 393.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Efficient protease with alanine specificity but only little elastolytic activity.Specifications
| CELA3B | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P08861, Q9CQ52 | |
| 23436, 298567, 67868 | |
| Recombinant Human ELA3B protein, Val29-His270 (Accession #P08861). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 0910001F22Rik; 2310074F01Rik; AI504000; CBPP; Cela3b; cholesterol-binding pancreatic protease; chymotrypsin like elastase family member 3B; chymotrypsin-like elastase family member 3B; chymotrypsin-like elastase family, member 3B; Ela3; ELA3B; elastase 3, pancreatic; elastase 3B, pancreatic; elastase IIIB; Elastase-3B; fecal elastase 1; pancreatic elastase 1; pancreatic endopeptidase E; procarboxypeptidase A complex component III; procarboxypeptidase A-S6 subunit III; PROCPA-S6 III; proproteinase E; protease E; proteinase E | |
| CELA3B | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| CELA3B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title