Learn More
Celecoxib Carboxylic Acid Acyl-Beta-D-glucuronide, TRC
Product Code. 30366477
Modifier l'affichage
Click to view available options
Quantity:
1 mg
Conditionnement:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
30366477 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30366477 Fournisseur Toronto Research Chemicals Code fournisseur TRCC2510151MG
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Spécification
Chemical Name or Material Celecoxib Carboxylic Acid Acyl-beta-D-glucuronide
CAS 264236-79-5
Molecular Formula C23H20F3N3O10S
Quantity 1 mg
Synonym 1-[4-[1-[4-(Aminosulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoate] β-D-Glucopyranuronic αcid
InChI Formula InChI=1S/C23H20F3N3O10S/c24-23(25,26)15-9-14(29(28-15)12-5-7-13(8-6-12)40(27,36)37)10-1-3-11(4-2-10)21(35)39-22-18(32)16(30)17(31)19(38-22)20(33)34/h1-9,16-19,22,30-32H,(H,33,34)(H2,27,36,37)/t16-,17-,18+,19-,22-/m0/s1
SMILES NS(C1=CC=C(N2C(C3=CC=C(C(O[C@H]4[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O)=O)O4)=O)C=C3)=CC(C(F)(F)F)=N2)C=C1)(=O)=O
IUPAC Name (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[4-[2-(4-sulfamoylphenyl)-5-(trifluoromethyl)pyrazol-3-yl]benzoyl]oxyoxane-2-carboxylic acid
Molecular Weight (g/mol) 587.48