missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CELF6 Monoclonal Antibody (OTI5G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811938
This item is not returnable.
View return policy
Description
RNA-binding protein implicated in the regulation of pre-mRNA alternative splicing. Mediates exon inclusion and/or exclusion in pre-mRNA that are subject to tissue-specific and developmentally regulated alternative splicing. Specifically activates exon 5 inclusion of TNNT2 in a muscle-specific splicing enhancer (MSE)-dependent manner. Promotes also exon exclusion of INSR pre-mRNA.Specifications
| CELF6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q7TN33, Q96J87 | |
| CELF6 | |
| Full length human recombit protein of human CELF6 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5G3 | |
| Unconjugated | |
| CELF6 | |
| BRUNOL6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 60677, 76183 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction