Learn More
CELF6 Monoclonal Antibody (OTI5G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811938S
This item is not returnable.
View return policy
Description
CELF6 Monoclonal antibody specifically detects CELF6 in Human samples. It is validated for Western BlotSpecifications
| CELF6 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96J87 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 60677 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5G3 | |
| Unconjugated | |
| CELF6 | |
| 6330569O16Rik; Bruno -like 6, RNA binding protein; BRUNOL6; bruno-like 6, RNA binding protein; bruno-like protein 6; CELF6; CELF-6; CUG-BP and ETR-3 like factor 6; CUG-BP- and ETR-3-like factor 6; CUGBP Elav-like family member 6; CUGBP, Elav-like family member 6; RNA-binding protein BRUNOL-6 | |
| Full length human recombinant protein of human CELF6 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |