Learn More
Lonza™ Cell Line Optimization 4D-Nucleofector™ X Kit
Product Code. 13409339
Click to view available options
No. of Reactions:
24
96
Unit Size:
24 reactions
96 reactions
Product Code. 13409339
Brand: Lonza™ V4XP9096
Specifications
Content And Storage 4°C
Vector pmaxGFP™
For Use With (Equipment) 4D-Nucleofector™ X Unit
Includes Includes 20μL Transfection volume, 0.675mL Nucleofector Solution, 4 x 0.15mL Supplement, 2x 50μg pmaxGFP Vector, 2 Single Nucleocuvette (100 μl), Store at 4°C, For Use with 4D-Nucleofector X Unit
No. of Reactions 96
Product Type Cell Line Optimization 4D-Nucleofector X Kit
Quantity 1 Each