missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CellAdhere™ Vitronectin, Human, Solution

Product Code. 17170002
Encompass_Preferred
Change view
Click to view available options
Quantity:
0.2 mL
Unit Size:
0.20mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17170002 0.2 mL 0.20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17170002 Supplier STEMCELL Technologies Supplier No. 07004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purified human vitronectin solution for tissue engineering research, cell culture, and biochemistry

  • Vitronectin is a common glycoprotein that is abundant in animal serum, and can also be found in the extracellular matrix deposited by many cell types
  • It is primarily used in cell culture as a surface coating to promote cell attachment and spreading
  • CellAdhere™ Vitronectin, Human, Solution is purified from human serum by the method of Hayman et al.) using an anti-Vitronectin monoclonal antibody affinity column and sterilized by 0.2 μm filtration
  • Vitronectin has a purity of > 95% based on Coomassie brilliant blue stain of 7.5% SDS-PAGE
  • Product is dissolved in 0.2 mL of 0.15 M NaCl, 0.005 M HEPES buffer at approximately pH 7.4
  • Final concentration of Vitronectin is 0.5 mg/mL
  • Fibronectin contamination is less than 0.04% based on immunoblotting
  • The optimal protein concentration may vary depending on the cell type being used, and therefore must be titrated for best results

Specifications

Quantity 0.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.