Learn More
Corning™ CellCube™ Modules
Shop All Corning Products
Click to view available options
Capacity (Metric):
600 mL
1.5 L
Tray Layers:
10
25
Surface Treatment:
CellBIND
Tissue Culture-treated
Unit Size:
Each
Pack of 2
Description
CellCube Modules provide:
- a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
- polystyrene growth surface optimally treated for the growth of attachment dependent cells.
- a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
- an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
Specifications
Specifications
| Description | Individually Bagged |
| Capacity (Metric) | 1.5 L |
| No. per Case | 1 |
| No. per Pack | 1 |
| Product Type | CellCube™ Module |
| Shape | Square |
| Graduated | No |
| Sterility | Sterile |
| Culture Area | 21,250 cm2 |
| Surface Treatment | Tissue Culture-treated |
| Show More |