Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, U-Shaped-Bottom Microplate
Color:Clear; Sterility:Sterile; Lid:With Lid
Brand: Greiner Bio-One 650180
Additional Details : Weight : 4.89600kg
Description
- Improved cell adhesion through physical surface treatment
- Compatible with automated systems
- Alphanumeric well coding
- High clarity of the microplates for optimal microscopic examinations
- Stackable
- Consecutive lot numbering
- Uniform footprint
- Can be pipetted precisely
- Suitable for +/- analyses
- Solid bottom Well profile: V-bottom (conical bottom)
- Growth area per well: 28 mm²
- Working volume per well: 40 μl - 200 μl
- With lid that enables gas exchange with minimal evaporation
- TC surface treatment Sterile Individually wrapped in peel-off packs for user convenience
Specifications
DIN ISO 9001 | |
Standard | |
0.28 mm2 | |
Clear | |
96 | |
Sterile | |
234 μL | |
Yes | |
4800 x g (swinging bucket rotor) | |
-20°C to +60°C |
RNase/DNase, human DNA free | |
With Lid | |
8 x 12 | |
Polystyrene | |
40 to 200 μL | |
Cell Culture | |
Round Bottom (U-well) | |
-20°C to +60°C | |
100/Cs. |