Learn More
Greiner Bio-One CELLSTAR TC treated Cell Culture Dish with vents
Description
- High-quality virgin polystyrene construction
- Nonpyrogenic
- Free of RNase and DNase
- Stackable
- Uniform gas exchange and reduced condensation
- Applications
- For optimal cell attachment
Specifications
Specifications
| Certifications/Compliance | Free of detectable DNase/RNase, human DNA and pyrogens. Contents non-cytotoxic |
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 100 mL |
| Color | Clear |
| Culture Area | 58 cm² |
| Material | Polystyrene |
| Shape | Round |
| Show More |