Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow are recommended for detection of yeasts and molds, particle count, and microscopy.
€ 141.00 - € 910.00
Specifications
Color | Gray/White Grid |
---|---|
Material | Cellulose Nitrate (Cellulose Mixed Ester) |
Pore Size | 0.45 μm |
Sterility | Sterile |
Grade | 130 |
Product Code | Brand | Diameter (Metric) | No. per Pack | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | No. per Pack | Price | Quantity & Availability | ||||
15922470
|
Sartorius Stedim Biotech™
130H6--47----ACN |
47 mm | 100/pk. |
€ 141.00
Pack of 100 |
Estimated Shipment: 08-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15932470
|
Sartorius Stedim Biotech™
130H6--47----ACR |
47 mm | 1000/pk. |
€ 900.00
Pack of 1000 |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15952470
|
Sartorius Stedim Biotech™
130H6--50----ACR |
50 mm | 1000/pk. |
€ 910.00
Pack of 1000 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15942470
|
Sartorius Stedim Biotech™
130H6--50----ACN |
50 mm | 100/pk. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Sterile, individually packaged filters
- Save preparatory time
- Perfect grid printing
- Filter colors allow the best contrast to the colonies or particles
- Outstanding microorganism recovery rate
- High flow membranes
- Sterilized by gamma irradiation
- Compatible with aqueous solutions (pH 4-8), hydrocarbons and several other organic solvents
Specifications
Gray/White Grid | |
0.45 μm | |
130 |
Cellulose Nitrate (Cellulose Mixed Ester) | |
Sterile | |
115 to 145 μm |