missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CELSR2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98104UV
This item is not returnable.
View return policy
Description
CELSR2 Polyclonal specifically detects CELSR2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CELSR2 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| cadherin EGF LAG seven-pass G-type receptor 2, Cadherin family member 10, cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila), CDHF10FLJ34118, EGFL2FLJ45143, EGF-like protein 2, EGF-like-domain, multiple 2, epidermal growth factor-like 2, Epidermal growth factor-like protein 2, Flamingo homolog 3, Flamingo1, FLJ45845, KIAA0279FLJ42737, MEGF3cadherin, EGF LAG seven-pass G-type receptor 2, flamingo (Drosophila) homolog, Multiple EGF-like domains protein 3, multiple epidermal growth factor-like domains 3, Multiple epidermal growth factor-like domains protein 3 | |
| Produced in rabbits immunized with E. coli-derived Human CELSR2 fragment. | |
| 0.1 mL | |
| GPCR | |
| 1952 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction