missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CELSR2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585908

Product Code. 16352815

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CELSR2 Polyclonal Antibody for Western Blot, ICC/IF

The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CELSR2
Polyclonal
Unconjugated
CELSR2
ADGRC2; adhesion G protein-coupled receptor C2; cadherin EGF LAG seven-pas G-type receptor 2; cadherin EGF LAG seven-pass G-type receptor 2; Cadherin family member 10; cadherin, EGF LAG seven-pass G-type receptor 2; cadherin, EGF LAG seven-pass G-type receptor 2 (flamingo homolog, Drosophila); CDHF10; Celsr2; EGFL2; EGF-like protein 2; EGF-like-domain, multiple 2; epidermal growth factor-like 2; epidermal growth factor-like protein 2; flamingo; Flamingo homolog; flamingo homolog 3; Flamingo1; flamingo-like protein celsr2; FLJ34118; FLJ42737; FLJ45143; FLJ45845; KIAA0279; MEGF3; mfmi1; mKIAA0279; Multiple EGF-like domains protein 3; multiple epidermal growth factor-like domains 3; multiple epidermal growth factor-like domains protein 3; seven-pass transmembrane cadherin
Rabbit
Antigen Affinity Chromatography
RUO
1952, 53883
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Q9HCU4, Q9R0M0
CELSR2
Synthetic peptide within Human Cadherin EGF LAG seven-pass G-type receptor 2 aa 21-70.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.