missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEMIP/KIAA1199 Antibody (V98P4E1*B7), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50336B
This item is not returnable.
View return policy
Description
CEMIP/KIAA1199 Monoclonal specifically detects CEMIP/KIAA1199 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| KIAA1199 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CEMIP | |
| Peptide Sequence - VTLDTEDHKA peptide immunogen is identical in equine (Equus caballus), 90% identical in mouse (Mus musculus) and rat (Rattus norvegicus) | |
| 0.1 mL | |
| Cancer | |
| 57214 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| V98P4E1*B7 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CCSP1, colon cancer secreted protein 1, IR2155535, KIAA1199, TMEM2L | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction