Learn More
Cenicriviroc Mesylate, MedChemExpress
Product Code. 16498827
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Product Code. 16498827
Brand: Medchem Express HY14882A
MedChemExpress Cenicriviroc Mesylate (TAK-652 Mesylate) is a dual CCR2/CCR5 antagonist, also inhibits both HIV-1 and HIV-2, and displays potent anti-inflammatory and antiinfective activity.
Cenicriviroc Mesylate (TAK-652 Mesylate) is a dual CCR2/CCR5 antagonist, also inhibits both HIV-1 and HIV-2, and displays potent anti-inflammatory and antiinfective activity.
Specifications
Chemical Name or Material Cenicriviroc Mesylate
CAS 497223-28-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C42H56N4O7S2
Quantity 5 mg
Synonym TAK-652 Mesylate TBR-652 Mesylate
Solubility Information DMSO : 100 mg/mL (126.10 mM; Need ultrasonic)
SMILES O=C(/C1=C/C2=CC(C3=CC=C(OCCOCCCC)C=C3)=CC=C2N(CC(C)C)CCC1)NC4=CC=C([S@](CC5=CN=CN5CCC)=O)C=C4.OS(=O)(C)=O
Molecular Weight (g/mol) 793.05
Formula Weight 793.05
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only