missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cenicriviroc Mesylate, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
Description
Cenicriviroc Mesylate (TAK-652 Mesylate) is a dual CCR2/CCR5 antagonist, also inhibits both HIV-1 and HIV-2, and displays potent anti-inflammatory and antiinfective activity.
Specifications
Specifications
| Chemical Name or Material | Cenicriviroc Mesylate |
| CAS | 497223-28-6 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C42H56N4O7S2 |
| Quantity | 10 mg |
| Synonym | TAK-652 Mesylate TBR-652 Mesylate |
| Solubility Information | DMSO : 100 mg/mL (126.10 mM; Need ultrasonic) |
| SMILES | O=C(/C1=C/C2=CC(C3=CC=C(OCCOCCCC)C=C3)=CC=C2N(CC(C)C)CCC1)NC4=CC=C([S@](CC5=CN=CN5CCC)=O)C=C4.OS(=O)(C)=O |
| Molecular Weight (g/mol) | 793.05 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction