missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPA Antibody (5A7-2E11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12892UV
This item is not returnable.
View return policy
Description
CENPA Monoclonal antibody specifically detects CENPA in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CENPA | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| 5A7-2E11 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Centromere autoantigen A, centromere protein A (17kD), centromere protein ACENP-Acentromere protein A, 17kDa, histone H3-like centromeric protein A | |
| Synthetic peptide corresponding to a portion of human CENP-A | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 1058 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction