Learn More
Invitrogen™ CENPA Monoclonal Antibody (5A7-2E11), PE
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545300
Product Code. 17848042
  • € 436.00 / 100µg
Description
1 μg/mL of MA5-45300 was sufficient for detection of CENPA in 20 μg of U2OS cell lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 18kDa. Recognizes Human CENP-A.
Centromeres are the differentiated chromosomal domains that specify the mitotic behavior of chromosomes. CENPA encodes a centromere protein which contains a histone H3 related histone fold domain that is required for targeting to the centromere. CENPA is proposed to be a component of a modified nucleosome or nucleosome-like structure in which it replaces 1 or both copies of conventional histone H3 in the (H3-H4)2 tetrameric core of the nucleosome particle.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CENPA
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative
P49450
Cenpa
Synthetic peptide corresponding to a portion of human CENP-A.
100 μg
Primary
Human
Antibody
IgG1
Western Blot, Immunocytochemistry
5A7-2E11
PE
Cenpa
CenH3; CENP A; CENPA; CENP-A; Centromere autoantigen A; centromere protein A; centromere protein A, 17kDa; centromere-specific histone; ggCENP-A; histone H3-like centromeric protein A; RGD1563607
Mouse
Protein G
RUO
1058
4°C