missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPB Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58103
This item is not returnable.
View return policy
Description
CENPB Polyclonal specifically detects CENPB in Human samples. It is validated for Western Blot.
Specifications
| CENPB | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CENP-B, centromere autoantigen B, Centromere protein B, centromere protein B (80kD), centromere protein B, 80kDa, major centromere autoantigen B | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 92%; Rat: 92%; Canine: 85%; Equine: 85%; Pig: 85%; Sheep: 85%. | |
| Human, Mouse, Rat, Pig, Bovine, Equine, Sheep, Yeast, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P07199 | |
| CENPB | |
| Synthetic peptides corresponding to CENPB(centromere protein B, 80kDa) The peptide sequence was selected from the C terminal of CENPB. Peptide sequence GGEDSDSDSEEEDDEEEDDEDEDDDDDEEDGDEVPVPSFGEAMAYFAMVK. | |
| 100 μL | |
| Cell Cycle and Replication, Chromatin Research, DNA Repair | |
| 1059 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction