missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPE, Mouse anti-Human, Clone: 1H12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB1924.100ug
This item is not returnable.
View return policy
Specifications
| CENPE | |
| Monoclonal | |
| Unconjugated | |
| In 20 mM KH2PO4, 150 mM NaCl, pH 7.2 (0.01% sodium azide) | |
| KIF10 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 1H12 | |
| Western Blot (1:500-1:2000) Immunoprecipitation (1:200) Immunofluorescence (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| centromere protein E, 312kDa | |
| CENPE | |
| Recombinant protein corresponding to full length human CENPE. | |
| RUO | |
| 1062 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction