missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPE Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144699
This item is not returnable.
View return policy
Description
Immunogen sequence: EKIENLTRML VTSSSLTLQQ ELKAKRKRRV TWCLGKINKM KNSNYADQFN IPTNITTKTH KLSINLLREI DESVCSESDV FSNTLDTLSE IEWNPATKLL NQENIESELN SLRADYDN Antigen sequence identity: Mouse 56%; Rat 60%.
14-3-3 sigma (stratifin, SFN, epithelial cell marker protein 1) is a member of a highly conserved family of 14-3-3 proteins that are present in all eukaryotic organisms. There are 7 known mammalian 14-3-3 isoforms epsilon, beta, zeta, gamma, theta, tau and they play important roles in many biological activities by binding to and altering the subcellular localization and/or stability of key molecules in various signaling cascades. 14-3-3 sigma was originally characterized as a human mammary epithelium marker and later rediscovered as an important molecule for cell cycle checkpoint regulation. Recently it has been reported that 14-3-3 sigma may serve as a prognosis marker predicting survival of pancreatic cancer patient treatment. 14-3-3 sigmais the only 14-3-3 isoform induced by the tumor suppressor protein p53, in response to <FONT face=Symbol>g</FONT>-irradiation and other DNA-damaging agents. 14-3-3 sigmais a p53-regulated inhibitor of G2/M progression and acts as a tumor suppressor gene that is inactivated by methylation of its 5' CpG islands in epithelial tumor cells. Two isoforms of human 14-3-3 sigma are produced by alternative splicing.
Specifications
| CENPE | |
| Polyclonal | |
| Unconjugated | |
| CENPE | |
| 312kDa; AU019344; BC049989; C530022J18; Cenpe; CENP-E; centromere autoantigen E; Centromere autoantigen E (312kD); centromere protein E; centromere protein E, 312kDa; centromere-associated protein E; centromeric protein E; KIF10; kinesin 10; kinesin family member 10; kinesin superfamily protein 10; Kinesin-7; Kinesin-related protein CENPE; MCPH13; Motor domain of KIF10; N-7 kinesin; PPP1R61; protein phosphatase 1, regulatory subunit 61 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1062 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q02224 | |
| CENPE | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 392-509. Recombinant protein control fragment (Product #RP-89483). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction