missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPH Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122101
This item is not returnable.
View return policy
Description
CENPH Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Centromere and kinetochore proteins play a critical role in centromere structure, kinetochore formation, and sister chromatid separation. The protein encoded by this gene colocalizes with inner kinetochore plate proteins CENP-A and CENP-C in both interphase and metaphase. It localizes outside of centromeric heterochromatin, where CENP-B is localized, and inside the kinetochore corona, where CENP-E is localized during prometaphase. It is thought that this protein can bind to itself, as well as to CENP-A, CENP-B or CENP-C. Multimers of the protein localize constitutively to the inner kinetochore plate and play an important role in the organization and function of the active centromere-kinetochore complex.
Specifications
| CENPH | |
| Polyclonal | |
| Unconjugated | |
| CENPH | |
| 1700021I11Rik; 2410018A12Rik; 2610042E16Rik; 2810046K12Rik; AU044255; CENPH; CENP-H; centromere autoantigen H; centromere protein H; Enp; ICEN35; Interphase centromere complex protein 35; kinetochore protein CENP-H; LOC681185; similar to centromere autoantigen H | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 26886, 64946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9H3R5, Q9QYM8 | |
| CENPH | |
| Recombinant protein Centromere Protein H. The antigen corresponds to amino acid range 28-241 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction