missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CENPJ Monoclonal Antibody (OTI3G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811852
This item is not returnable.
View return policy
Description
CENPJ Monoclonal antibody specifically detects CENPJ in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CENPJ | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O35800, Q16665, Q61221 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 15251, 29560, 3091 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI3G8 | |
| Unconjugated | |
| HIF1A | |
| 4932437H03Rik; BM032; Cenpj; CENP-J; Centromere protein J; centrosomal P4.1-associated protein; CPAP; Gm81; LAG-3-associated protein; LAP; LIP1; LYST-interacting protein 1; LYST-interacting protein LIP1; LYST-interacting protein LIP7; MCPH6; microcephaly; Sas4; Sas-4; SASS4; SCKL4 | |
| Human recombinant protein fragment corresponding to amino acids 531-826 of human HIF1a (NP_001521) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction