missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CENPK Polyclonal Antibody
GREENER_CHOICE

Product Code. 13290529
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13290529 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13290529 Supplier Invitrogen™ Supplier No. PA526324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CENPK Polyclonal Antibody for Western Blot, IHC (P)

CENPK is a subunit of a CENPH -CENPI -associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CENPK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CENPK
Gene Accession No. Q9BS16, Q9ESN5
Gene Alias AF5alpha; B130045K24Rik; BB232189; C530004N04Rik; CENPK; Cenp-K; Centromere protein K; FKSG14; ICEN37; Interphase centromere complex protein 37; leucine zipper protein FKSG14; p33; Protein AF-5alpha; Solt; Solzt; SoxLZ/Sox6 leucine zipper binding protein in testis; SoxLZ/Sox6 leucine zipper-binding protein in testis; SoxLZ/Sox6-binding protein Solt
Gene Symbols CENPK
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 239-266 amino acids from the C-terminal region of human CENPK.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60411, 64105
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.