missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CENPK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597579
This item is not returnable.
View return policy
Description
CENPK Polyclonal Antibody for Western Blot, IHC (P), ELISA
CENPK is a subunit of a CENPH -CENPI -associated centromeric complex that targets CENPA to centromeres and is required for proper kinetochore function and mitotic progression.
Specifications
| CENPK | |
| Polyclonal | |
| Unconjugated | |
| CENPK | |
| AF5alpha; B130045K24Rik; BB232189; C530004N04Rik; CENPK; Cenp-K; Centromere protein K; FKSG14; ICEN37; Interphase centromere complex protein 37; leucine zipper protein FKSG14; p33; Protein AF-5alpha; Solt; Solzt; SoxLZ/Sox6 leucine zipper binding protein in testis; SoxLZ/Sox6 leucine zipper-binding protein in testis; SoxLZ/Sox6-binding protein Solt | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64105 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BS16 | |
| CENPK | |
| Recombinant Human Centromere protein K protein (1-269AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction